5-Amino-2,2-difluoro-1,3-benzodioxole ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline

A181733-1G A181733-250MG A181733-25G A181733-5G
ALA181733-1GEA 50.26 USD
ALA181733-1G ALA181733250MG ALA181733-25G ALA181733-5G
5-Amino-2,2-difluoro-1,3-benzodioxole ≥98%
5-Amino-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 119
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00190144
CAS Number: 1544-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d26229f3c703737696cc56da277e440601ecb4f0&vid=d26229f3c703737696cc56da277e440601ecb4f0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR