2-Aminothiophene-3-carboxamide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A181429-1G A181429-25G
ALA181429-1GEA 26.51 USD
ALA181429-1G ALA181429-25G
2-Aminothiophene-3-carboxamide ≥98%
2-Aminothiophene-3-carboxamide
Formula: C₅H₆N₂OS
Melting Pt: 154.0 - 158.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00706455
CAS Number: 14080-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91c5b8bebcd383d1ad5ca36e649463c60f98dfd1&vid=91c5b8bebcd383d1ad5ca36e649463c60f98dfd1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR