(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Aminomethylphenylboronic acid, pinacol ester

A181385-1G A181385-5G
ALA181385-1GEA 77.81 USD
ALA181385-1G ALA181385-5G
(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine ≥96%
(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine
Formula: C₁₃H₂₀BNO₂
MW: 233.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06213229
CAS Number: 138500-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ac10b13be45abb8b8e98558ea69227976b4a330&vid=8ac10b13be45abb8b8e98558ea69227976b4a330&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR