2-Amino-4,6-dichloropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A179785-1G A179785-25G A179785-5G
ALA179785-1GEA 16.06 USD
ALA179785-1G ALA179785-25G ALA179785-5G
2-Amino-4,6-dichloropyridine ≥97%
2-Amino-4,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD04037218
CAS Number: 116632-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee154a3f5ffef3f667fe903fe531f41f49a4494d&vid=ee154a3f5ffef3f667fe903fe531f41f49a4494d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR