5'-Bromo-2'-methylacetanilide ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Acetamido-4-bromotoluene, N-(5-Bromo-2-methylphenyl)acetamide

A179775-1G A179775-25G A179775-5G
ALA179775-1GEA 37.91 USD
ALA179775-1G ALA179775-25G ALA179775-5G
5'-Bromo-2'-methylacetanilide ≥98%
5'-Bromo-2'-methylacetanilide
Formula: C₉H₁₀BrNO
MW: 228.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520466
CAS Number: 116436-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f67e2c83e4ccb0d67e2eaa8e50b015ae9c09267&vid=5f67e2c83e4ccb0d67e2eaa8e50b015ae9c09267&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR