(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Aminomethy-4-(4-fluorobenzyl)morpholine

A179484-1G A179484-250MG A179484-25G A179484-5G
ALA179484-1GEA 19.86 USD
ALA179484-1G ALA179484250MG ALA179484-25G ALA179484-5G
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine ≥98%
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine
Formula: C₁₂H₁₇FN₂O
MW: 224.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658145
CAS Number: 112914-13-3
EINECS: 601-212-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3c3d2ccf32f1eead3a6113f79e9153316cbf21e&vid=f3c3d2ccf32f1eead3a6113f79e9153316cbf21e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR