2-Amino-4-bromobenzamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A179453-1G A179453-250MG
ALA179453-1GEA 132.91 USD
ALA179453-1G ALA179453250MG
2-Amino-4-bromobenzamide ≥97%
2-Amino-4-bromobenzamide
Formula: C₇H₇BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD12674809
CAS Number: 112253-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a468997b202007c80ccf2b1143d7ad5e1dfef5d&vid=3a468997b202007c80ccf2b1143d7ad5e1dfef5d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR