5-(Trifluoromethyl)anthranilic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A177547-100G A177547-1G A177547-250MG A177547-25G A177547-5G
ALA177547-100GEA 25461.81 USD
ALA177547-100G ALA177547-1G ALA177547250MG ALA177547-25G ALA177547-5G
5-(Trifluoromethyl)anthranilic acid ≥97%
5-(Trifluoromethyl)anthranilic acid
Formula: C₈H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01569537
CAS Number: 83265-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aed487c919083a55e0de6017712c3361abfbf3b2&vid=aed487c919083a55e0de6017712c3361abfbf3b2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR