3-Azabicyclo[3.1.0]hexane hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-azabicyclo[3.1.0]hexane hydrochloride

A177313-1G A177313-250MG A177313-50MG A177313-5G
ALA177313-1GEA 36.01 USD
ALA177313-1G ALA177313250MG ALA177313-50MG ALA177313-5G
3-Azabicyclo[3.1.0]hexane hydrochloride ≥97%
3-Azabicyclo[3.1.0]hexane hydrochloride
Formula: C₅H₉N·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD12197361
CAS Number: 73799-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed5d7bee493c0349011e5b7124995904d73892c6&vid=ed5d7bee493c0349011e5b7124995904d73892c6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR