3-Amino-1H-pyrazol-5-ol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-amino-1H-pyrazol-5-ol

A176721-5G A176721-100G A176721-1G A176721-25G
ALA176721-5GEA 18.91 USD
ALA176721-5G ALA176721-100G ALA176721-1G ALA176721-25G
3-Amino-1H-pyrazol-5-ol ≥97%
3-Amino-1H-pyrazol-5-ol
Formula: C₃H₅N₃O
MW: 99.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00233932
CAS Number: 53666-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44d61fb26050be1ef8ce67d283dfac5535297a1c&vid=44d61fb26050be1ef8ce67d283dfac5535297a1c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR