PRIMA-1MET ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A176692-100MG A176692-25MG A176692-5MG
ALA176692100MGEA 617.41 USD
ALA176692100MG ALA176692-25MG ALA176692-5MG
PRIMA-1MET ≥97%
PRIMA-1MET

PRIMA-1Met restores wild-type conformation and function to mutant p53, and triggers apoptosis in tumor cells. PRIMA-1Met also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance.

Formula: C₁₀H₁₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD16294187
CAS Number: 5291-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a01574cc408f32d296f7faae77904673fd47c3&vid=86a01574cc408f32d296f7faae77904673fd47c3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR