3-Amino-2,2-dimethylpropionamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Amino-2,2-dimethylpropanamide

A176132-100G A176132-1G A176132-250MG A176132-25G A176132-5G
ALA176132-100GEA 664.91 USD
ALA176132-100G ALA176132-1G ALA176132250MG ALA176132-25G ALA176132-5G
3-Amino-2,2-dimethylpropionamide ≥97%
3-Amino-2,2-dimethylpropionamide
Formula: C5H12N2O
Storage Temperature: Refrigerator
MDL Number: MFCD06797568
CAS Number: 324763-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5eb235e1e717588e74677f57670d16eeea48fe1c&vid=5eb235e1e717588e74677f57670d16eeea48fe1c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR