AN2/AVex-73 hydrochloride salt ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: AE-37 hydrochloride, AVex-73 hydrochloride, Blarcamesine hydrochloride

A175437-250MG A175437-25MG A175437-50MG A175437-5MG A175437-100MG
ALA175437250MGEA 2140.26 USD
ALA175437250MG ALA175437-25MG ALA175437-50MG ALA175437-5MG ALA175437100MG
AN2/AVex-73 hydrochloride salt ≥97%
AN2/AVex-73 hydrochloride salt

Product Introduction:Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM

Formula: C₁₉H₂₄ClNO
Storage Temperature: Freezer
CAS Number: 195615-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19a6985932a575bfed88ec7ac9e4c678cf680a08&vid=19a6985932a575bfed88ec7ac9e4c678cf680a08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR