7-Aza-spiro[3.5]nonane hydrochloride 97
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
7-Azaspiro[3.5]nonane hydrochloride
A174040-100MG
A174040-1G
A174040-250MG
ALA174040100MGEA
36.96
USD
ALA174040100MG
ALA174040-1G
ALA174040250MG
7-Aza-spiro[3.5]nonane hydrochloride 97
7-Aza-spiro[3.5]nonane hydrochloride
Formula:
C₈H₁₅N·ClH MW: 161.672 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD23105809 CAS Number: 1414885-16-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...