(1-Aminocyclobutyl)methanol hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (1-aminocyclobutyl)methanol hydrochloride

A173832-100MG A173832-1G A173832-250MG A173832-5G
ALA173832100MGEA 16.06 USD
ALA173832100MG ALA173832-1G ALA173832250MG ALA173832-5G
(1-Aminocyclobutyl)methanol hydrochloride ≥97%
(1-Aminocyclobutyl)methanol hydrochloride
Formula: C₅H₁₁NO·ClH
MW: 137.608 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015024
CAS Number: 1392213-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6f684cbd7430b42b7ecaf3f671f9c5f2132ebf7&vid=c6f684cbd7430b42b7ecaf3f671f9c5f2132ebf7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR