3-Amino-2-bromophenol ≥97%
Supplier: ALADDIN SCIENTIFIC
Warning
|
Synonyms:
3-amino-2-bromophenol
A171600-100MG
A171600-1G
A171600-250MG
A171600-5G
ALA171600100MGEA
10.36
USD
ALA171600100MG
ALA171600-1G
ALA171600250MG
ALA171600-5G
3-Amino-2-bromophenol ≥97%
3-Amino-2-bromophenol
Formula:
C₆H₆BrNO MW: 188.022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846453 CAS Number: 100367-36-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...