4-Amino-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-2-methylphenylboronic acid, pinacol ester, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A171258-1G A171258-250MG A171258-5G
ALA171258-1GEA 190.86 USD
ALA171258-1G ALA171258250MG ALA171258-5G
4-Amino-2-methylphenylboronic acid pinacol ester ≥97%
4-Amino-2-methylphenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₂
Storage Temperature: Ambient
MDL Number: MFCD08458198
CAS Number: 631911-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c3464c8ef5216888a329b3b6a201c3a189faa7&vid=51c3464c8ef5216888a329b3b6a201c3a189faa7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR