1-(2-Aminophenyl)pyrrole ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-(1H-Pyrrol-1-yl)aniline
A171139-1G
A171139-200MG
A171139-250MG
A171139-25G
A171139-5G
ALA171139-1GEA
23.66
USD
ALA171139-1G
ALA171139200MG
ALA171139250MG
ALA171139-25G
ALA171139-5G
1-(2-Aminophenyl)pyrrole ≥95%
1-(2-Aminophenyl)pyrrole
1-(2-Aminophenyl)pyrrole participates in Pt(IV)-catalyzed hydroamination triggered cyclization reaction to yield fused pyrrolo [1,2-a] quinoxalines. It reacts with aromatic or heteroaromatic aldehydes in ethanol and catalytic amounts of acetic acid to yield 4,5-dihydropyrrolo[1,2-a]quinoxalines. Thin films of poly(1-(2-aminophenyl)pyrrole) has been prepared via oxidative electropolymerization.1-(2-Aminophenyl)pyrrole was used in the synthesis of 4-substituted pyrrolo[1,2-a]quinoxaline derivatives.
Formula:
C₁₀H₁₀N₂ MW: 158.20 g/mol Melting Pt: 96-98°C Storage Temperature: Ambient |
MDL Number:
MFCD00005344 CAS Number: 6025-60-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...