1-Acetylindole ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
N-Acetylindole, Indol-1-yl methyl ketone, 1-(1-Indolyl)-1-ethanone, 1-(1H-Indol-1-yl)ethanone
A171066-5G
A171066-1G
ALA171066-5GEA
242.16
USD
ALA171066-5G
ALA171066-1G
1-Acetylindole ≥98%
1-Acetylindole
Formula:
C₁₀H₉NO MW: 159.19 Density: 1.387 Flash Pt: >113 °C |
MDL Number:
MFCD00038005 CAS Number: 576-15-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...