6-Aminophthalide ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 6-Aminoisobenzofuran-1(3H)-one, 6-AMINOPHTHALIDE

A171047-2.5G A171047-500MG
ALA171047-2.5GEA 621.21 USD
ALA171047-2.5G ALA171047500MG
6-Aminophthalide ≥95%
6-Aminophthalide
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 182-185°C
Storage Temperature: Ambient
MDL Number: MFCD00033530
CAS Number: 57319-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=830f106bd77c2bf0d34be678792edcd7861c1480&vid=830f106bd77c2bf0d34be678792edcd7861c1480&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR