3-Amino-4-phenyl-1H-pyrazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Phenyl-1H-pyrazol-3-amine

A170987-1G A170987-5G
ALA170987-1GEA 46.46 USD
ALA170987-1G ALA170987-5G
3-Amino-4-phenyl-1H-pyrazole ≥97%
3-Amino-4-phenyl-1H-pyrazole
Formula: C₉H₉N₃
MW: 159.19 g/mol
Melting Pt: 176-180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01693715
CAS Number: 5591-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7c297b530cf6fd86e78419e8d24cd835d3ae6f2&vid=c7c297b530cf6fd86e78419e8d24cd835d3ae6f2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR