5-(Methylthio)-1,3,4-thiadiazol-2-amine ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5-(methylthio)-1,3,4-thiadiazole

A170827-1G A170827-25G A170827-5G A170827-100G
ALA170827-1GEA 17.96 USD
ALA170827-1G ALA170827-25G ALA170827-5G ALA170827-100G
5-(Methylthio)-1,3,4-thiadiazol-2-amine ≥99%
5-(Methylthio)-1,3,4-thiadiazol-2-amine

Product Introduction:2-Amino-5-(methylthio)-1,3,4-thiadiazole is a 2-amino-5-substituted-1,3,4-thiadiazole. Its density at 25°C is 1.4020g/ml.Application:2-Amino-5-(methylthio)-1,3,4-thiadiazole may be used in the synthesis of 9-methylthio-7,7-dioxo-7,7a-dihydro-5-oxo-7λ6,10-dithia-8,11-diaza-cyclopenta[b] phenantren-6-one by reacting with 4-chloro-2-oxo-2H-chromene-3-sulfonyl chloride.

Formula: C₃H₅N₃S₂
MW: 147.22 g/mol
Melting Pt: 178-181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00175752
CAS Number: 5319-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f240b7f63d915b389305c8f31dc9a46df70e3ec9&vid=f240b7f63d915b389305c8f31dc9a46df70e3ec9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR