2-Amino-4,5-dimethyl-3-furonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-4,5-dimethylfuran-3-carbonitrile, 2-Amino-4,5-dimethyl-3-furancarbonitrile

A170692-1G A170692-5G
ALA170692-1GEA 56.91 USD
ALA170692-1G ALA170692-5G
2-Amino-4,5-dimethyl-3-furonitrile ≥95%
2-Amino-4,5-dimethyl-3-furonitrile
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 163-169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274264
CAS Number: 5117-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58e977bd545104d0864f15e9174ec2f2dcbc5a7b&vid=58e977bd545104d0864f15e9174ec2f2dcbc5a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR