2-Acetyl-7-methoxybenzofuran ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-(7-Methoxy-1-benzofuran-2-yl)-1-ethanone, 1-(7-Methoxybenzofuran-2-yl)ethanone

A170336-1G A170336-5G
ALA170336-1GEA 36.96 USD
ALA170336-1G ALA170336-5G
2-Acetyl-7-methoxybenzofuran ≥98%
2-Acetyl-7-methoxybenzofuran
Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Boiling Pt: 180 °C
Melting Pt: 92-96°C
MDL Number: MFCD00016911
CAS Number: 43071-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa8523511c9940e1c2cf84de3e5c16d9b71d6c1d&vid=aa8523511c9940e1c2cf84de3e5c16d9b71d6c1d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR