2-Amino-5-methylbenzyl Alcohol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5-methylbenzyl alcohol

A169771-5G A169771-1G A169771-250MG A169771-25G
ALA169771-5GEA 443.56 USD
ALA169771-5G ALA169771-1G ALA169771250MG ALA169771-25G
2-Amino-5-methylbenzyl Alcohol ≥97%
2-Amino-5-methylbenzyl Alcohol
Formula: C₈H₁₁NO
MW: 137.17 g/mol
Melting Pt: 123-126°C
Storage Temperature: Ambient
MDL Number: MFCD00074995
CAS Number: 34897-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4f45c134c338cd1f2e1eab36e53d3311dc9d6d2&vid=a4f45c134c338cd1f2e1eab36e53d3311dc9d6d2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR