4-Aminoindan ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,3-Dihydro-1H-inden-4-amine, 4-Aminoindane

A169551-1G A169551-25G A169551-5G
ALA169551-1GEA 9.41 USD
ALA169551-1G ALA169551-25G ALA169551-5G
4-Aminoindan ≥97%
4-Aminoindan

4-Aminoindan undergoes nucleophlic addition reaction with 2-chloroacetylisocyanate to yield 1-(2-chloroacetyl)-3-(2,3-dihydro-1H-inden-4-yl)-urea.

Formula: C₉H₁₁N
MW: 133.19
Boiling Pt: 65
Melting Pt: -2
Storage Temperature: Ambient
MDL Number: MFCD00082598
CAS Number: 32202-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84f07e35973e0ff3f39d16690d4650a0f9ec6949&vid=84f07e35973e0ff3f39d16690d4650a0f9ec6949&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR