2-(2-Aminophenyl)benzothiazole ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A169345-5G
A169345-1G
A169345-250MG
ALA169345-5GEA
96.81
USD
ALA169345-5G
ALA169345-1G
ALA169345250MG
2-(2-Aminophenyl)benzothiazole ≥97%
2-(2-Aminophenyl)benzothiazole
Formula:
C₁₃H₁₀N₂S Melting Pt: 126-130°C Storage Temperature: Ambient |
MDL Number:
MFCD00937701 CAS Number: 29483-73-6 UN: 2811 ADR: 6.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...