3-Amino-4-chlorobenzenesulfonamide ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A169318-100MG A169318-1G A169318-250MG A169318-5G
ALA169318100MGEA 34.11 USD
ALA169318100MG ALA169318-1G ALA169318250MG ALA169318-5G
3-Amino-4-chlorobenzenesulfonamide ≥95%
3-Amino-4-chlorobenzenesulphonamide
Formula: C₆H₇ClN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD01165741
CAS Number: 29092-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45fd8541b72b7a482c69f6974b467213ea119a6d&vid=45fd8541b72b7a482c69f6974b467213ea119a6d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR