3-Aminophthalimide ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
4-Aminoisoindoline-1,3-dione
A169034-10G
A169034-1G
A169034-25G
A169034-5G
ALA169034-10GEA
55.01
USD
ALA169034-10G
ALA169034-1G
ALA169034-25G
ALA169034-5G
3-Aminophthalimide ≥97%
3-Aminophthalimide
Formula:
C₈H₆N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00041853 CAS Number: 2518-24-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...