Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-indan-2-carboxylic acid methyl ester hydrochloride

A168328-100MG A168328-1G A168328-250MG A168328-5G
ALA168328100MGEA 9.41 USD
ALA168328100MG ALA168328-1G ALA168328250MG ALA168328-5G
Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride ≥95%
Methyl 2-amino-2,3-dihydro-1H-indene-2-carboxylate hydrochloride
Formula: C₁₁H₁₄ClNO₂
MW: 227.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437619
CAS Number: 199330-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b23272702d571064ae48b720bb388a491b4d363&vid=7b23272702d571064ae48b720bb388a491b4d363&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR