6-Amino-3-cyano-2-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile

A168088-1G A168088-250MG A168088-25G A168088-5G
ALA168088-1GEA 49.31 USD
ALA168088-1G ALA168088250MG ALA168088-25G ALA168088-5G
6-Amino-3-cyano-2-methylpyridine ≥97%
6-Amino-3-cyano-2-methylpyridine

6-Amino-3-cyano-2-methylpyridine is a useful nitrile building block for proteomics research and synthesis.

Formula: C₇H₇N₃
Melting Pt: 162-166 °C
Storage Temperature: Ambient
MDL Number: MFCD05865116
CAS Number: 183428-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79bffcb47496167ee7dba6f6a23367c8cfd937f6&vid=79bffcb47496167ee7dba6f6a23367c8cfd937f6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR