2-Amino-4-methylthiazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Methylthiazol-2-ylamine, 4-Methylthiazol-2-amine, 2-Amino-4-methylthiazol

A167666-100G A167666-25G A167666-5G
ALA167666-100GEA 104.41 USD
ALA167666-100G ALA167666-25G ALA167666-5G
2-Amino-4-methylthiazole ≥98%
2-Amino-4-methylthiazole
Formula: C₄H₆N₂S
Boiling Pt: 232 °C
Melting Pt: 45 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005329
CAS Number: 1603-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1f10ede0445c8eb52058b2012083b6c76d3b4&vid=f6b1f10ede0445c8eb52058b2012083b6c76d3b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR