(3-Amino-4-pyridinyl)methanol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Aminopyridine-4-methanol, (3-Amino-pyridin-4-yl)-methanol, 3-Amino-4-hydroxymethylpyridine

A167487-1G A167487-250MG A167487-50MG
ALA167487-1GEA 127.21 USD
ALA167487-1G ALA167487250MG ALA167487-50MG
(3-Amino-4-pyridinyl)methanol ≥95%
(3-Amino-4-pyridinyl)methanol
Formula: C₆H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD06658330
CAS Number: 152398-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2920a84cbe87e17ad8a149f90f737cb157014dc3&vid=2920a84cbe87e17ad8a149f90f737cb157014dc3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR