2-Acetylamino-5-bromo-6-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Acetamido-5-bromo-6-methylpyridine, 6-Acetamido-3-bromo-2-methylpyridine

A167256-1G A167256-25G A167256-5G
ALA167256-1GEA 41.71 USD
ALA167256-1G ALA167256-25G ALA167256-5G
2-Acetylamino-5-bromo-6-methylpyridine ≥97%
2-Acetylamino-5-bromo-6-methylpyridine

2-Acetylamino-5-bromo-6-methylpuridine is used to prepare palladium-catalyzed cross-coupling reactions of pyridylboronic acids with heteroaryl halides.

Formula: C₈H₉BrN₂O
MW: 229.0700 g/mol
Melting Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD03909755
CAS Number: 142404-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7903fbb474d272f24a9560bcc830dda098e07b08&vid=7903fbb474d272f24a9560bcc830dda098e07b08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR