(4-Aminopyridin-3-yl)methanol ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Amino-3-pyridinemethanol, (4-Amino-pyridin-3-yl)-methanol

A167140-250MG A167140-5G A167140-1G
ALA167140250MGEA 49.31 USD
ALA167140250MG ALA167140-5G ALA167140-1G
(4-Aminopyridin-3-yl)methanol ≥97%
(4-Aminopyridin-3-yl)methanol
Formula: C₆H₈N₂O
Melting Pt: 167-169°C
Storage Temperature: Refrigerator
MDL Number: MFCD06203066
CAS Number: 138116-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59c84844865324f5da8fe6088ea4857368cb1294&vid=59c84844865324f5da8fe6088ea4857368cb1294&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR