3-Aminopropylphosphonic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A166928-1G A166928-250MG A166928-5G
ALA166928-1GEA 211.76 USD
ALA166928-1G ALA166928250MG ALA166928-5G
3-Aminopropylphosphonic acid ≥98%
3-Aminopropylphosphonic acid

3-Aminopropylphosphonic acid was used as a selective antagonist of GABA(C) receptor.

Formula: C3H10NO3P
Melting Pt: 294°C
Storage Temperature: Ambient
MDL Number: MFCD00008222
CAS Number: 13138-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc881622a200d3a546884f0415cfc8dd8a9513ad&vid=cc881622a200d3a546884f0415cfc8dd8a9513ad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR