7-Amino-2,4-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 7-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one

A166881-1G A166881-250MG A166881-50MG
ALA166881-1GEA 250.71 USD
ALA166881-1G ALA166881250MG ALA166881-50MG
7-Amino-2,4-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one ≥98%
7-Amino-2,4-dimethyl-2H-benzo[b][1,4]oxazin-3(4H)-one
Formula: C₁₀H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09907443
CAS Number: 130137-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47f7fcc4fa7dc6712aa3d6ccf57d28449d9a28ef&vid=47f7fcc4fa7dc6712aa3d6ccf57d28449d9a28ef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR