(±)-2-Aminoheptane ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Aminoheptane, (±)-Tuaminoheptane, 1-Methylhexylamine, 2-Heptylamine, (±)-1-Methylhexylamine, Tuaminoheptane

A166657-5G A166657-25G
ALA166657-5GEA 63.56 USD
ALA166657-5G ALA166657-25G
(±)-2-Aminoheptane ≥99%
(±)-2-Aminoheptane
Formula: C₇H₁₇N
MW: 115.22 g/mol
Boiling Pt: 143
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00008101
CAS Number: 123-82-0
Merck Index: 12,09934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63096aaef7fa82cae74f461e4844eace8b75acbe&vid=63096aaef7fa82cae74f461e4844eace8b75acbe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR