2-Amino-5-fluoronicotinic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A166354-250MG A166354-50MG
ALA166354250MGEA 93.96 USD
ALA166354250MG ALA166354-50MG
2-Amino-5-fluoronicotinic acid ≥97%
2-Amino-5-fluoronicotinic acid
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190373
CAS Number: 1196154-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19f34006d6dd28f5d2ec772bbc6c96d939660e9f&vid=19f34006d6dd28f5d2ec772bbc6c96d939660e9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR