2-Amino-3-bromophenol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A166175-1G A166175-250MG A166175-5G
ALA166175-1GEA 52.16 USD
ALA166175-1G ALA166175250MG ALA166175-5G
2-Amino-3-bromophenol ≥97%
2-Amino-3-bromophenol
Formula: C₆H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD08445676
CAS Number: 116435-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e81d072ae61445438704b5b56a66f21f2c3b9c55&vid=e81d072ae61445438704b5b56a66f21f2c3b9c55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR