Nα,Nα-Bis(carboxymethyl)-L-lysine hydrate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: L-Lysine, N2,N2-bis(carboxymethyl)-, hydrate (1:x), (S)-N-(5-Amino-1-carboxypentyl)iminodiacetic acid hydrate

A166003-25G A166003-5G A166003-10G A166003-1G A166003-250MG
ALA166003-25GEA 4145.71 USD
ALA166003-25G ALA166003-5G ALA166003-10G ALA166003-1G ALA166003250MG
Nα,Nα-Bis(carboxymethyl)-L-lysine hydrate ≥97%
Nα,Nα-Bis(carboxymethyl)-L-lysine hydrate
Formula: C₁₀H₂₀N₂O₇
MW: 280.27 g/mol
Melting Pt: 219 °C
Storage Temperature: Ambient
MDL Number: MFCD24387945
CAS Number: 941689-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc348a464014beacbd4ba47ce4022407a8dbb8f8&vid=dc348a464014beacbd4ba47ce4022407a8dbb8f8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR