4-Amino-N-methylbenzenemethanesulfonamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Amino-N-methyl-alpha-toluenesulfonamide, 4-Amino-N-methyl-α-toluenesulphonamide, 4-Amino-N-methyl-α-toluenesulfonamide, 1-(4-Aminophenyl)-N-methylmethanesulphonamide, 4-Amino-N-methylbenzenemethanesulphonamide, 4-Amino-N-methyl-alpha-toluenesulphonamide, 4-Amino-N-methylbenzenemethanesulfonamide, 1-(4-Aminophenyl)-N-methylmethanesulfonamide

A165897-1G A165897-25G A165897-5G
ALA165897-1GEA 44.56 USD
ALA165897-1G ALA165897-25G ALA165897-5G
4-Amino-N-methylbenzenemethanesulfonamide ≥97%
4-Amino-N-methylbenzenemethanesulfonamide
Formula: C₈H₁₂N₂O₂S
Melting Pt: 140-142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671697
CAS Number: 109903-35-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=468df69abf8e3602da5936a27af264a957948b62&vid=468df69abf8e3602da5936a27af264a957948b62&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR