1-Amino-1-cyclopentanemethanol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A165541-10G A165541-1G A165541-250MG A165541-5G
ALA165541-10GEA 682.01 USD
ALA165541-10G ALA165541-1G ALA165541250MG ALA165541-5G
1-Amino-1-cyclopentanemethanol ≥97%
1-Amino-1-cyclopentanemethanol

Application:1-Amino-1-cyclopentanemethanol was used in the synthesis of xanthine-derived heterocyclic fused systems. It was also used as building block in medicinal chemistry synthesis.

Formula: C₆H₁₃NO
Boiling Pt: 85~90℃
Melting Pt: 20°C
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010491
CAS Number: 10316-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f43832935baf5b1f85c8b7d389855a87e4f99b4&vid=6f43832935baf5b1f85c8b7d389855a87e4f99b4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR