4-Aminopyridine-2-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A165325-1G A165325-250MG A165325-25G A165325-5G
ALA165325-1GEA 185.16 USD
ALA165325-1G ALA165325250MG ALA165325-25G ALA165325-5G
4-Aminopyridine-2-carboxylic acid ≥97%
4-Aminopyridine-2-carboxylic acid
Formula: C₆H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02685671
CAS Number: 100047-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f87b41bb3cd251f8da03565891cfc42a2b687443&vid=f87b41bb3cd251f8da03565891cfc42a2b687443&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR