H-D-4-Pal-OH·2HCl ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: (R)-2-Amino-3-(4-pyridyl)propionic acid hydrochlorate, 3-(4-Pyridyl)-D-alanine hydrochlorate, 3-(4-Pyridyl)-D-alanine·2HCl, H-D-4-Pal-OH.2HCl, D-4-Pyridylalanine hydrochlorate

A165279-1G A165279-25G A165279-5G
ALA165279-1GEA 27.46 USD
ALA165279-1G ALA165279-25G ALA165279-5G
H-D-4-Pal-OH·2HCl ≥98%
H-D-4-Pal-OH·2HCl
Formula: C₈H₁₀N₂O₂·₂HCl
MW: 239.2 g/mol
MDL Number: MFCD09842075
CAS Number: 174096-41-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eafce5819958ad55ec1bcdfe903b4cfa9933aff&vid=4eafce5819958ad55ec1bcdfe903b4cfa9933aff&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR