Azido PEG amine

Supplier: ALADDIN SCIENTIFIC

Synonyms: N3-PEG-NH2

A163282-100MG A163282-500MG
ALA163282100MGEA 112.01 USD
ALA163282100MG ALA163282500MG
Azido PEG amine
Azido PEG amine

Azide/azido functionalized polyethylene glycol (N3-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials. Azide group can react with alkyne in aqueous solution catalyzed by copper. It can also be reduced into amine group easily while another functional group can be react with other molecules or functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.

Formula: NH₂(CH₂CH₂O)nH
Storage Temperature: Freezer
CAS Number: 32130-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR