Amino PEG Amine
Supplier: ALADDIN SCIENTIFIC
Synonyms: NH2-PEG-NH2
A163238-100MG A163238-25MG
ALA163238100MGEA 431.21 USD
ALA163238100MG ALA163238-25MG
Amino PEG Amine
Amino PEG Amine
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.