Alkyne PEG thiol
Supplier: ALADDIN SCIENTIFIC
A163215-100MG A163215-500MG
ALA163215100MGEA 326.71 USD
ALA163215100MG ALA163215500MG
Alkyne PEG thiol
Alkyne PEG thiol
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.