Alkyne PEG Biotin
Supplier: ALADDIN SCIENTIFIC
Synonyms: ALK-PEG-Biotin
A163209-100MG A163209-500MG
ALA163209100MGEA 262.11 USD
ALA163209100MG ALA163209500MG
Alkyne PEG Biotin
Alkyne PEG Biotin
Alkyne functionalized polyethylene glycol (Alk-PEG-X) is a bifunctional PEG derivative that can be used to modify proteins, peptides and other materials via Click Chemistry. Alkyne group can react with azide in aqueous solution catalyzed by copper. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.