Aldehyde PEG Aldehyde
Supplier: ALADDIN SCIENTIFIC
Synonyms: CHO-PEG-CHO
A163187-100MG A163187-500MG
ALA163187100MGEA 150.01 USD
ALA163187100MG ALA163187500MG
Aldehyde PEG Aldehyde
Aldehyde PEG Aldehyde
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.